2-Methoxy-3-(trifluormethyl)-5-pyridinboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 2-Methoxy-3-(trifluoromethyl)pyridine-5-boronic Acid Pinacol Ester

PN-5712-1G
COBBPN-5712-1GEA 681 EUR
COBBPN-5712-1G
2-Methoxy-3-(trifluormethyl)-5-pyridinboronsäurepinakolester
2-Methoxy-3-(trifluormethyl)-5-pyridinboronsäurepinakolester
Formel: C₁₃H₁₇BF₃NO₃
Molecular Weight: 303,0900 g/mol
MDL: MFCD12026094
CAS-Nummer: 1150561-61-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR