2-Nitro-5-(trifluormethyl)phenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

PN-5754-1G
COBBPN-5754-1GEA 0 EUR
COBBPN-5754-1G
2-Nitro-5-(trifluormethyl)phenylboronsäurepinakolester
2-Nitro-5-(trifluormethyl)phenylboronsäurepinakolester
Formel: C₁₃H₁₅BF₃NO₄
MDL: MFCD12756467
CAS-Nummer: 1218791-26-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR