5-Cyan-2-nitrophenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

PN-5755-1G
COBBPN-5755-1GEA 595 EUR
COBBPN-5755-1G
5-Cyan-2-nitrophenylboronsäurepinakolester
5-Cyan-2-nitrophenylboronsäurepinakolester
Formel: C₁₃H₁₅BN₂O₄
MDL: MFCD12546471
CAS-Nummer: 1218791-27-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR