2-Cyan-5-nitrophenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

PN-5756-1G PN-5756-250M
COBBPN-5756-1GEA 0 EUR
COBBPN-5756-1G COBBPN-5756-250M
2-Cyan-5-nitrophenylboronsäurepinakolester
2-Cyan-5-nitrophenylboronsäurepinakolester
Formel: C₁₃H₁₅BN₂O₄
MDL: MFCD12546472
CAS-Nummer: 1218791-28-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR