2,6-Difluor-(4-methoxycarbonyl)phenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 2,6-Difluoro-4-(methoxycarbonyl)benzeneboronic acid pinacol ester, Methyl 3,5-difluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate

PN-5841-1G PN-5841-250M
COBBPN-5841-1GEA 0 EUR
COBBPN-5841-1G COBBPN-5841-250M
2,6-Difluor-(4-methoxycarbonyl)phenylboronsäurepinakolester
2,6-Difluor-(4-methoxycarbonyl)phenylboronsäurepinakolester
Formel: C₁₄H₁₇BF₂O₄
MDL: MFCD12546491
CAS-Nummer: 1218791-32-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR