4-(Trifluormethylthio)phenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

PN-5864-1G PN-5864-250M
COBBPN-5864-1GEA 0 EUR
COBBPN-5864-1G COBBPN-5864-250M
4-(Trifluormethylthio)phenylboronsäurepinakolester
4-(Trifluormethylthio)phenylboronsäurepinakolester
Formel: C₁₃H₁₆BF₃O₂S
MDL: MFCD11042448
CAS-Nummer: 1005206-25-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR