2-(tert-Butylamino)-5-pyrimidinboronsäurepinakolester

Lieferant: COMBI-BLOCKS

PN-5905-1G
COBBPN-5905-1GEA 341 EUR
COBBPN-5905-1G
2-(tert-Butylamino)-5-pyrimidinboronsäurepinakolester
2-(tert-Butylamino)-5-pyrimidinboronsäurepinakolester
Formel: C₁₄H₂₄BN₃O₂
MDL: MFCD12546547
CAS-Nummer: 1218791-43-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR