2-(Butylamino)-5-pyrimidinboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: N-Butyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidin-2-amine

PN-5925-1G
COBBPN-5925-1GEA 336 EUR
COBBPN-5925-1G
2-(Butylamino)-5-pyrimidinboronsäurepinakolester
2-(Butylamino)-5-pyrimidinboronsäurepinakolester
Formel: C₁₄H₂₄BN₃O₂
Molecular Weight: 277,1700 g/mol
MDL: MFCD12546558
CAS-Nummer: 1218791-48-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR