2-(2-Methoxyethylamino)-5-pyrimidinboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: N-(2-Methoxyethyl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidin-2-amine

PN-5927-1G
COBBPN-5927-1GEA 451 EUR
COBBPN-5927-1G
2-(2-Methoxyethylamino)-5-pyrimidinboronsäurepinakolester
2-(2-Methoxyethylamino)-5-pyrimidinboronsäurepinakolester
Formel: C₁₃H₂₂BN₃O₃
Molecular Weight: 279,143 g/mol
MDL: MFCD12546559
CAS-Nummer: 1202805-24-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR