2-(3-Methoxypropylamino)-5-pyrimidinboronsäurepinakolester

Lieferant: COMBI-BLOCKS

PN-5928-1G
COBBPN-5928-1GEA 404 EUR
COBBPN-5928-1G
2-(3-Methoxypropylamino)-5-pyrimidinboronsäurepinakolester
2-(3-Methoxypropylamino)-5-pyrimidinboronsäurepinakolester
Formel: C₁₄H₂₄BN₃O₃
MDL: MFCD12546560
CAS-Nummer: 1218789-31-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR