2-(Cyclohexylamino)-5-pyrimidinboronsäurepinakolester

Lieferant: COMBI-BLOCKS

PN-5934-1G
COBBPN-5934-1GEA 404 EUR
COBBPN-5934-1G
2-(Cyclohexylamino)-5-pyrimidinboronsäurepinakolester
2-(Cyclohexylamino)-5-pyrimidinboronsäurepinakolester
Formel: C₁₆H₂₆BN₃O₂
MDL: MFCD12546564
CAS-Nummer: 1218789-35-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR