2-(Phenylamino)-5-pyrimidinboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: N-Phenyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidin-2-amine

PN-5935-1G
COBBPN-5935-1GEA 595 EUR
COBBPN-5935-1G
2-(Phenylamino)-5-pyrimidinboronsäurepinakolester
2-(Phenylamino)-5-pyrimidinboronsäurepinakolester
Formel: C₁₆H₂₀BN₃O₂
Molecular Weight: 297,159 g/mol
MDL: MFCD12546565
CAS-Nummer: 956128-05-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR