3-Chlor-5-ethoxyphenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

PN-5954-1G
COBBPN-5954-1GEA 497 EUR
COBBPN-5954-1G
3-Chlor-5-ethoxyphenylboronsäurepinakolester
3-Chlor-5-ethoxyphenylboronsäurepinakolester
Formel: C₁₄H₂₀BClO₃
MDL: MFCD12546577
CAS-Nummer: 1218789-40-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR