3-Brom-5-(tert-butyldimethylsilyloxy)phenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: (3-Bromo-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenoxy)(tert-butyl)dimethylsilane

PN-5970-1G
COBBPN-5970-1GEA 595 EUR
COBBPN-5970-1G
3-Brom-5-(tert-butyldimethylsilyloxy)phenylboronsäurepinakolester
3-Brom-5-(tert-butyldimethylsilyloxy)phenylboronsäurepinakolester
Formel: C₁₈H₃₀BBrO₃Si
Molecular Weight: 413,2300 g/mol
MDL: MFCD12546591
CAS-Nummer: 1218789-51-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR