3-Butoxy-5-(methoxycarbonyl)phenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

PN-5978-1G
COBBPN-5978-1GEA 303 EUR
COBBPN-5978-1G
3-Butoxy-5-(methoxycarbonyl)phenylboronsäurepinakolester
3-Butoxy-5-(methoxycarbonyl)phenylboronsäurepinakolester
Formel: C₁₈H₂₇BO₅
MDL: MFCD12546598
CAS-Nummer: 1218789-66-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR