3-(tert-Butyldimethylsilyloxy)-5-methylphenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

PN-5987-1G
COBBPN-5987-1GEA 864 EUR
COBBPN-5987-1G
3-(tert-Butyldimethylsilyloxy)-5-methylphenylboronsäurepinakolester
3-(tert-Butyldimethylsilyloxy)-5-methylphenylboronsäurepinakolester
Formel: C₁₉H₃₃BO₃Si
MDL: MFCD12546607
CAS-Nummer: 1218789-84-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR