1,3-Dimethyl-1H-pyrazol-5-boronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 1,3-Dimethyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole

PN-6021-1G
COBBPN-6021-1GEA 262 EUR
COBBPN-6021-1G
1,3-Dimethyl-1H-pyrazol-5-boronsäurepinakolester
1,3-Dimethyl-1H-pyrazol-5-boronsäurepinakolester
Formel: C₁₁H₁₉BN₂O₂
Molecular Weight: 222,0918 g/mol
MDL: MFCD05664574
CAS-Nummer: 847818-79-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR