2-Amino-4-(trifluormethoxy)phenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

PN-6028-1G
COBBPN-6028-1GEA 864 EUR
COBBPN-6028-1G
2-Amino-4-(trifluormethoxy)phenylboronsäurepinakolester
2-Amino-4-(trifluormethoxy)phenylboronsäurepinakolester
Formel: C₁₃H₁₇BF₃NO₃
MDL: MFCD16618968
CAS-Nummer: 1256359-27-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR