2-(Bis-Boc-amino)-5-pyrimidinboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: tert-Butyl (tert-butoxycarbonyl)(5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidin-2-yl)carbamate

PN-6038-1G
COBBPN-6038-1GEA 169 EUR
COBBPN-6038-1G
2-(Bis-Boc-amino)-5-pyrimidinboronsäurepinakolester
2-(Bis-Boc-amino)-5-pyrimidinboronsäurepinakolester
Formel: C₂₀H₃₂BN₃O₆
Molecular Weight: 421,2956 g/mol
MDL: MFCD12913985
CAS-Nummer: 1190423-36-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR