2-(4-Methoxybenzylamino)-5-pyrimidinboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: N-(4-Methoxybenzyl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidin-2-amine

PN-6045-1G
COBBPN-6045-1GEA 595 EUR
COBBPN-6045-1G
2-(4-Methoxybenzylamino)-5-pyrimidinboronsäurepinakolester
2-(4-Methoxybenzylamino)-5-pyrimidinboronsäurepinakolester
Formel: C₁₈H₂₄BN₃O₃
Molecular Weight: 341,2100 g/mol
MDL: MFCD15143596
CAS-Nummer: 1218790-01-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR