4-Chlor-5-cyan-2-fluorphenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

PN-6078-1G
COBBPN-6078-1GEA 623 EUR
COBBPN-6078-1G
4-Chlor-5-cyan-2-fluorphenylboronsäurepinakolester
4-Chlor-5-cyan-2-fluorphenylboronsäurepinakolester
Formel: C₁₃H₁₄BClFNO₂
MDL: MFCD13195765
CAS-Nummer: 1218790-13-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR