Propen-1-ylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

PN-6132-1G
COBBPN-6132-1GEA 266 EUR
COBBPN-6132-1G
Propen-1-ylboronsäurepinakolester
Propen-1-ylboronsäurepinakolester
Formel: C₉H₁₇BO₂
MDL: MFCD15143598
CAS-Nummer: 72824-05-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR