3,5-Dichlor-4-fluorphenylboronsäurepinakolester
Lieferant: COMBI-BLOCKS
Synonyme:
2-(3,5-Dichloro-4-fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
PN-6145-1G
COBBPN-6145-1GEA
595
EUR
COBBPN-6145-1G
3,5-Dichlor-4-fluorphenylboronsäurepinakolester
3,5-Dichlor-4-fluorphenylboronsäurepinakolester
Formel:
C₁₂H₁₄BCl₂FO₂ Molecular Weight: 290,96 g/mol |
MDL:
MFCD15143599 CAS-Nummer: 1092485-88-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...