1-Boc-6-methyl-3-indolboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: tert-Butyl 6-methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole-1-carboxylate

PN-6188-1G
COBBPN-6188-1GEA 303 EUR
COBBPN-6188-1G
1-Boc-6-methyl-3-indolboronsäurepinakolester
1-Boc-6-methyl-3-indolboronsäurepinakolester
Formel: C₂₀H₂₈BNO₄
Molecular Weight: 357,2500 g/mol
MDL: MFCD16295108
CAS-Nummer: 1256359-86-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR