1-Boc-5-cyan-3-indolboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: tert-Butyl 5-cyano-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole-1-carboxylate

PN-6215-1G
COBBPN-6215-1GEA 742 EUR
COBBPN-6215-1G
1-Boc-5-cyan-3-indolboronsäurepinakolester
1-Boc-5-cyan-3-indolboronsäurepinakolester
Formel: C₂₀H₂₅BN₂O₄
Molecular Weight: 368.2345 g/mol
MDL: MFCD15143604
CAS-Nummer: 1185427-07-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR