4-(Methoxycarbonyl)furan-2-boronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: Methyl 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)furan-3-carboxylate

PN-6294-1G
COBBPN-6294-1GEA 623 EUR
COBBPN-6294-1G
4-(Methoxycarbonyl)furan-2-boronsäurepinakolester
4-(Methoxycarbonyl)furan-2-boronsäurepinakolester
Formel: C₁₂H₁₇BO₅
Molecular Weight: 252,0700 g/mol
MDL: MFCD12407248
CAS-Nummer: 1111096-29-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR