1-(2-Tetrahydropyranyl)-1H-pyrazol-3,5-diboronsäurepinakolester

Lieferant: COMBI-BLOCKS

PN-6471-1G
COBBPN-6471-1GEA 1490 EUR
COBBPN-6471-1G
1-(2-Tetrahydropyranyl)-1H-pyrazol-3,5-diboronsäurepinakolester
1-(2-Tetrahydropyranyl)-1H-pyrazol-3,5-diboronsäurepinakolester
Formel: C₂₀H₃₄B₂N₂O₅
MDL: MFCD16660311
CAS-Nummer: 1256360-29-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR