1H-Pyrazol-3-boronsäurepinakolester

Lieferant: COMBI-BLOCKS

PN-8344-1G
COBBPN-8344-1GEA 153 EUR
COBBPN-8344-1G
1H-Pyrazol-3-boronsäurepinakolester
1H-Pyrazol-3-boronsäurepinakolester
Formel: C₉H₁₅BN₂O₂
MDL: MFCD07368044
CAS-Nummer: 844501-71-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR