3-Amino-4-fluorphenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 3-Amino-4-fluorobenzeneboronic acid pinacol ester, 2-Fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline, 3-Amino-4-fluorophenylboronic acid, pinacol ester

PN-8365-1G
COBBPN-8365-1GEA 225 EUR
COBBPN-8365-1G
3-Amino-4-fluorphenylboronsäurepinakolester
3-Amino-4-fluorphenylboronsäurepinakolester
Formel: C₁₂H₁₇BFNO₂
Molecular Weight: 237,08 g/mol
MDL: MFCD11044430
CAS-Nummer: 1003575-43-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR