3,4,5-Trichlorphenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 3,4,5-Trichlorophenylboronic acid,pinacol ester, 4,4,5,5-Tetramethyl-2-(3,4,5-trichlorophenyl)-1,3,2-dioxaborolane

PN-8385-1G
COBBPN-8385-1GEA 277 EUR
COBBPN-8385-1G
3,4,5-Trichlorphenylboronsäurepinakolester
3,4,5-Trichlorphenylboronsäurepinakolester
Formel: C₁₂H₁₄BCl₃O₂
Molecular Weight: 307,41 g/mol
MDL: MFCD11504973
CAS-Nummer: 942069-95-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR