Tetrahydropyran-4-boronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 4,4,5,5-Tetramethyl-2-(tetrahydro-2H-pyran-4-yl)-1,3,2-dioxaborolane

PN-8522-1G
COBBPN-8522-1GEA 285 EUR
COBBPN-8522-1G
Tetrahydropyran-4-boronsäurepinakolester
Tetrahydropyran-4-boronsäurepinakolester
Formel: C₁₁H₂₁BO₃
Molecular Weight: 212,0936 g/mol
MDL: MFCD11506065
CAS-Nummer: 1131912-76-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR