3-Chlor-5-(trifluormethyl)phenylboronsäurepinakolester
Lieferant: COMBI-BLOCKS
Synonyme:
3-Chloro-5-(trifluoromethyl)benzeneboronic acid pinacol ester, 2-(3-Chloro-5-(trifluoromethyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
PN-8529-1G
COBBPN-8529-1GEA
149
EUR
COBBPN-8529-1G
3-Chlor-5-(trifluormethyl)phenylboronsäurepinakolester
3-Chlor-5-(trifluormethyl)phenylboronsäurepinakolester
Formel:
C₁₃H₁₅BClF₃O₂ Molecular Weight: 306,52 g/mol |
MDL:
MFCD11856039 CAS-Nummer: 942069-65-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...