4,5-Dimethoxy-2-(methoxycarbonyl)phenylboronsäurepinakolester
Lieferant: COMBI-BLOCKS
Synonyme:
Methyl 4,5-dimethoxy-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
PN-8533-1G
COBBPN-8533-1GEA
486
EUR
COBBPN-8533-1G
4,5-Dimethoxy-2-(methoxycarbonyl)phenylboronsäurepinakolester
4,5-Dimethoxy-2-(methoxycarbonyl)phenylboronsäurepinakolester
Formel:
C₁₆H₂₃BO₆ Molecular Weight: 322,1600 g/mol |
MDL:
MFCD16036165 CAS-Nummer: 1201566-80-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...