2-(tert-Butoxycarbonyl)phenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

PN-8609-1G
COBBPN-8609-1GEA 0 EUR
COBBPN-8609-1G
2-(tert-Butoxycarbonyl)phenylboronsäurepinakolester
2-(tert-Butoxycarbonyl)phenylboronsäurepinakolester
Formel: C₁₇H₂₅BO₄
MDL: MFCD03453053
CAS-Nummer: 956229-69-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR