3,3-Diethoxy-1-propylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

PN-8612-250M
COBBPN-8612-250MEA 210 EUR
COBBPN-8612-250M
3,3-Diethoxy-1-propylboronsäurepinakolester
3,3-Diethoxy-1-propylboronsäurepinakolester
Formel: C₁₃H₂₇BO₄
Molecular Weight: 258,17 g/mol
MDL: MFCD03788723
CAS-Nummer: 165904-27-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR