2-Morpholino-5-pyridinboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 6-(4-Morpholinyl)pyridine-3-boronic acid pinacol ester, 2-(Morpholin-4-yl)pyridine-5-boronic acid pinacol ester, 4-(5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl)morpholine

PN-8632-1G
COBBPN-8632-1GEA 73.7 EUR
COBBPN-8632-1G
2-Morpholino-5-pyridinboronsäurepinakolester
2-Morpholino-5-pyridinboronsäurepinakolester
Formel: C₁₅H₂₃BN₂O₃
Molecular Weight: 290,17 g/mol
MDL: MFCD04039874
CAS-Nummer: 485799-04-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR