2,2'-Bithiophen-5-boronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 2-([2,2'-Bithiophen]-5-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

PN-8634-1G
COBBPN-8634-1GEA 89.2 EUR
COBBPN-8634-1G
2,2'-Bithiophen-5-boronsäurepinakolester
2,2'-Bithiophen-5-boronsäurepinakolester
Formel: C₁₄H₁₇BO₂S₂
Molecular Weight: 292,2 g/mol
MDL: MFCD04039974
CAS-Nummer: 479719-88-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR