1,4-Benzodioxan-6-boronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 1,4-Benzodioxane-6-boronic acid, pinacol ester, 2,3-Dihydro-1,4-benzodioxine-6-boronic acid, pinacol ester, 2-(2,3-Dihydrobenzo[b][1,4]dioxin-6-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

PN-8646-1G
COBBPN-8646-1GEA 141 EUR
COBBPN-8646-1G
1,4-Benzodioxan-6-boronsäurepinakolester
1,4-Benzodioxan-6-boronsäurepinakolester
Formel: C₁₄H₁₉BO₄
Molecular Weight: 262,1093 g/mol
MDL: MFCD05663890
CAS-Nummer: 517874-21-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR