3-(4-Methyl-1-piperazinylcarbonyl)phenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 3-(4-Methyl-1-piperazinylcarbonyl)benzeneboronic acid pinacol ester, (4-Methylpiperazin-1-yl)(3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)methanone

PN-8652-1G
COBBPN-8652-1GEA 125 EUR
COBBPN-8652-1G
3-(4-Methyl-1-piperazinylcarbonyl)phenylboronsäurepinakolester
3-(4-Methyl-1-piperazinylcarbonyl)phenylboronsäurepinakolester
Formel: C₁₈H₂₇BN₂O₃
Molecular Weight: 330,23 g/mol
MDL: MFCD05864308
CAS-Nummer: 883738-38-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR