4-Amino-3-methoxyphenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 4-Amino-3-methoxyphenylboronic acid, pinacol ester, 2-Methoxy-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline

PN-8671-1G
COBBPN-8671-1GEA 236 EUR
COBBPN-8671-1G
4-Amino-3-methoxyphenylboronsäurepinakolester
4-Amino-3-methoxyphenylboronsäurepinakolester
Formel: C₁₃H₂₀BNO₃
Molecular Weight: 249,12 g/mol
MDL: MFCD06795657
CAS-Nummer: 461699-81-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR