4-Amino-3-nitrophenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

PN-8679-5G PN-8679-1G
COBBPN-8679-5GEA 0 EUR
COBBPN-8679-5G COBBPN-8679-1G
4-Amino-3-nitrophenylboronsäurepinakolester
4-Amino-3-nitrophenylboronsäurepinakolester
Formel: C₁₂H₁₇BN₂O₄
MDL: MFCD06795680
CAS-Nummer: 833486-94-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR