2-(Cyclohexylamino)-5-pyridinboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: N-Cyclohexyl-5-(tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-amine

PN-8698-250M
COBBPN-8698-250MEA 208 EUR
COBBPN-8698-250M
2-(Cyclohexylamino)-5-pyridinboronsäurepinakolester
2-(Cyclohexylamino)-5-pyridinboronsäurepinakolester
Formel: C₁₇H₂₇BN₂O₂
MDL: MFCD06798277
CAS-Nummer: 1073354-34-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR