1-(2-Morpholinoethyl)-1H-pyrazol-4-boronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 4-(2-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazol-1-yl)ethyl)morpholine

PN-8727-1G
COBBPN-8727-1GEA 301 EUR
COBBPN-8727-1G
1-(2-Morpholinoethyl)-1H-pyrazol-4-boronsäurepinakolester
1-(2-Morpholinoethyl)-1H-pyrazol-4-boronsäurepinakolester
Formel: C₁₅H₂₆BN₃O₃
Molecular Weight: 307,1962 g/mol
MDL: MFCD08062342
CAS-Nummer: 864754-18-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR