2-Chlor-3-methyl-5-pyridinboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 2-Chloro-3-methylpyridine-5-boronic Acid Pinacol Ester

PN-8728-1G
COBBPN-8728-1GEA 204 EUR
COBBPN-8728-1G
2-Chlor-3-methyl-5-pyridinboronsäurepinakolester
2-Chlor-3-methyl-5-pyridinboronsäurepinakolester
Formel: C₁₂H₁₇BClNO₂
Molecular Weight: 253,54 g/mol
MDL: MFCD08063084
CAS-Nummer: 1010101-07-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR