2,6-Difluor-4-formylphenylboronsäurepinakolester
Lieferant: COMBI-BLOCKS
PN-8736-1G
COBBPN-8736-1GEA
0
EUR
COBBPN-8736-1G
2,6-Difluor-4-formylphenylboronsäurepinakolester
2,6-Difluor-4-formylphenylboronsäurepinakolester
Formel:
C₁₃H₁₅BF₂O₃ |
MDL:
MFCD08276848 CAS-Nummer: 870717-92-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...