1-(Ethoxycarbonylmethyl)-1H-pyrazol-4-boronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: Ethyl 2-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazol-1-yl)acetate

PN-8762-1G
COBBPN-8762-1GEA 210 EUR
COBBPN-8762-1G
1-(Ethoxycarbonylmethyl)-1H-pyrazol-4-boronsäurepinakolester
1-(Ethoxycarbonylmethyl)-1H-pyrazol-4-boronsäurepinakolester
Formel: C₁₃H₂₁BN₂O₄
Molecular Weight: 280,1278 g/mol
MDL: MFCD08706026
CAS-Nummer: 864754-16-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR