4,4-Dimethylcyclohex-2-en-1-on-2-ylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 4,4-Dimethyl-2-cyclohexen-1-one-2-boronic acid pinacol ester, 4,4-Dimethyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)cyclohex-2-enone

PN-8768-1G PN-8768-250M
COBBPN-8768-1GEA 0 EUR
COBBPN-8768-1G COBBPN-8768-250M
4,4-Dimethylcyclohex-2-en-1-on-2-ylboronsäurepinakolester
4,4-Dimethylcyclohex-2-en-1-on-2-ylboronsäurepinakolester
Formel: C₁₄H₂₃BO₃
MDL: MFCD09027074
CAS-Nummer: 219489-09-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR