2-Chlor-3-cyan-4-pyridinboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 2-Chloro-3-cyanopyridine-4-boronic acid pinacol ester

PN-8772-250M
COBBPN-8772-250MEA 462 EUR
COBBPN-8772-250M
2-Chlor-3-cyan-4-pyridinboronsäurepinakolester
2-Chlor-3-cyan-4-pyridinboronsäurepinakolester
Formel: C₁₂H₁₄BClN₂O₂
Molecular Weight: 264,52 g/mol
MDL: MFCD09037479
CAS-Nummer: 878194-94-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR