3-Cyan-4-pyridinboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 3-Cyanopyridine-4-boronic acid, pinacol ester

PN-8773-250M
COBBPN-8773-250MEA 154 EUR
COBBPN-8773-250M
3-Cyan-4-pyridinboronsäurepinakolester
3-Cyan-4-pyridinboronsäurepinakolester
Formel: C₁₂H₁₅BN₂O₂
Molecular Weight: 230,07 g/mol
MDL: MFCD09037481
CAS-Nummer: 878194-92-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR