6-(Boc-methylamino)-3-pyridinboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 6-(N-Boc-methylamino)pyridine-3-boronic acid pinacol ester, tert-Butyl methyl(5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl)carbamate

PN-8775-1G PN-8775-250M PN-8775-100M
COBBPN-8775-1GEA 0 EUR
COBBPN-8775-1G COBBPN-8775-250M COBBPN-8775-100M
6-(Boc-methylamino)-3-pyridinboronsäurepinakolester
6-(Boc-methylamino)-3-pyridinboronsäurepinakolester
Formel: C₁₇H₂₇BN₂O₄
MDL: MFCD09037484
CAS-Nummer: 1032758-87-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR